Anti-ATP synthase C Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB96655-50EA 853 EUR
ABCAAB96655-50
Anti-ATP synthase C Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ATP synthase C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR