Anti-PON2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96553-50EA 882 EUR
ABCAAB96553-50
Anti-PON2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PON2 (paraoxonase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR