Anti-KAT2B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96510-100EA 840 EUR
ABCAAB96510-100
Anti-KAT2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KAT2B (K(lysine) acetyltransferase 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR