Anti-CACNA1S Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB96413-50EA 840 EUR
ABCAAB96413-50
Anti-CACNA1S Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CACNA1S (calcium channel, voltage-dependent, L type, alpha 1S subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human