Anti-CAP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96354-50EA 859 EUR
ABCAAB96354-50
Anti-CAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAP1 (CAP, adenylate cyclase-associated protein 1 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR