Anti-BAIAP2L1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB96349-50EA 797 EUR
ABCAAB96349-50
Anti-BAIAP2L1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BAIAP2L1 (BAI1-associated protein 2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR