Anti-CBS Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96252-100EA 916 EUR
ABCAAB96252-100
Anti-CBS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR