Anti-DNAJB2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96095-50EA 797 EUR
ABCAAB96095-50
Anti-DNAJB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNAJB2 (DnaJ (Hsp40) homolog, subfamily B, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR