Anti-CD244 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 244F4]

Lieferant: Abcam
ABCAAB95807-50EA 476 EUR
ABCAAB95807-50
Anti-CD244 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 244F4]
Antikörper
Type: Primary
Antigen: CD244 (CD244 molecule, natural killer cell receptor 2B4)
Clonality: monoclonal
Clone: 244F4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR