Anti-CD8B Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: H35-17.2]

Lieferant: Abcam
ABCAAB95772-50EA 476 EUR
ABCAAB95772-50
Anti-CD8B Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: H35-17.2]
Antikörper
Type: Primary
Antigen: CD8B (CD8b molecule)
Clonality: monoclonal
Clone: H35-17.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR