Anti-CD8B Rat Monoclonal Antibody [clone: H35-17.2]

Lieferant: Abcam
ABCAAB95768-100EA 840 EUR
ABCAAB95768-100
Anti-CD8B Rat Monoclonal Antibody [clone: H35-17.2]
Antikörper
Type: Primary
Antigen: CD8B (CD8b molecule)
Clonality: monoclonal
Clone: H35-17.2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR