Anti-ITGA2B Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MWReg30]
Lieferant: Abcam
ABCAAB95726-50EA 852 EUR
ABCAAB95726-50
Anti-ITGA2B Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MWReg30]
Antikörper
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone: MWReg30
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse