Anti-ITGA6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: GoH3]
Lieferant: Abcam
ABCAAB95703-50EA 422 EUR
ABCAAB95703-50
Anti-ITGA6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: GoH3]
Antikörper
Type: Primary
Antigen: ITGA6 (integrin, alpha 6)
Clonality: monoclonal
Clone: GoH3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Bestellen