Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5E10]
Lieferant: Abcam
ABCAAB95700-50EA 423 EUR
ABCAAB95700-50
Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5E10]
Antikörper
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: 50000000000
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human