Anti-IFNGR1 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2E2]
Lieferant: Abcam
ABCAAB95673-50EA 834 EUR
ABCAAB95673-50
Anti-IFNGR1 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2E2]
Antikörper
Type: Primary
Antigen: IFNGR1 (interferon gamma receptor 1)
Clonality: monoclonal
Clone: 200
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse