Anti-CD8A Mouse monoclonal antibody FITC (Fluorescein) [clone: HIT8a]

Lieferant: Abcam
ABCAAB95591-50EA 379 EUR
ABCAAB95591-50
Anti-CD8A Mouse monoclonal antibody FITC (Fluorescein) [clone: HIT8a]
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone: HIT8a
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR