Anti-TUBGCP6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB95172-100EA 877 EUR
ABCAAB95172-100
Anti-TUBGCP6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TUBGCP6 (tubulin, gamma complex associated protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR