Anti-FAN1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB95171-100EA 859 EUR
ABCAAB95171-100
Anti-FAN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAN1 (FANCD2/FANCI-associated nuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR