Anti-Metapneumovirus Fusion Protein Mouse Monoclonal Antibody [clone: hMPV 24]
Lieferant: Abcam
ABCAAB94800-50EA 860 EUR
ABCAAB94800-50
Anti-Metapneumovirus Fusion Protein Mouse Monoclonal Antibody [clone: hMPV 24]
Antikörper
Type: Primary
Antigen: Metapneumovirus Fusion Protein
Clonality: monoclonal
Clone: hMPV 24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen