Anti-EMX2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB94713-100EA 844 EUR
ABCAAB94713-100
Anti-EMX2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EMX2 (empty spiracles homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR