Anti-LPL Mouse Monoclonal Antibody [clone: 5D2]
Lieferant: Abcam
ABCAAB93898-100EA 852 EUR
ABCAAB93898-100
Anti-LPL Mouse Monoclonal Antibody [clone: 5D2]
Antikörper
Type: Primary
Antigen: LPL (lipoprotein lipase)
Clonality: monoclonal
Clone: 5D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human