Anti-ARHGAP39 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB93780-100EA 835 EUR
ABCAAB93780-100
Anti-ARHGAP39 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARHGAP39 (Rho GTPase activating protein 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR