Anti-Ly76 Rat monoclonal antibody PE (Phycoerythrin) [clone: TER-119]

Lieferant: Abcam
ABCAAB93586-100EA 755 EUR
ABCAAB93586-100
Anti-Ly76 Rat monoclonal antibody PE (Phycoerythrin) [clone: TER-119]
Type: Primary
Antigen: Ly76
Clonality: monoclonal
Clone: TER-119
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR