Anti-IL3RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6H6]

Lieferant: Abcam
ABCAAB93523-50EA 834 EUR
ABCAAB93523-50
Anti-IL3RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6H6]
Antikörper
Type: Primary
Antigen: IL3RA (interleukin 3 receptor, alpha (low affinity))
Clonality: monoclonal
Clone: 6H6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR