Anti-CD8A Rabbit Monoclonal Antibody [clone: EP1150Y]

Lieferant: Abcam
ABCAAB93278-100EA 949 EUR
ABCAAB93278-100
Anti-CD8A Rabbit Monoclonal Antibody [clone: EP1150Y]
Antikörper
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone: EP1150Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR