Anti-NKX2-2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB93188-100EA 781 EUR
ABCAAB93188-100
Anti-NKX2-2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NKX2-2 (NK2 homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR