Anti-CCR6 Mouse Monoclonal Antibody [clone: MM0066-3L1]

Lieferant: Abcam
ABCAAB93086-100EA 859 EUR
ABCAAB93086-100
Anti-CCR6 Mouse Monoclonal Antibody [clone: MM0066-3L1]
Antikörper
Type: Primary
Antigen: CCR6 (chemokine (C-C motif) receptor 6)
Clonality: monoclonal
Clone: MM0066-3L1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR