Anti-Chitinase 3 like protein 3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB93034-100EA 860 EUR
ABCAAB93034-100
Anti-Chitinase 3 like protein 3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Chitinase 3 like protein 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR