Anti-BCAR1 Rabbit monoclonal antibody unconjugated [clone: EPR1860]

Lieferant: Abcam
ABCAAB92699-100EA 780 EUR
ABCAAB92699-100
Anti-BCAR1 Rabbit monoclonal antibody unconjugated [clone: EPR1860]
Antikörper
Type: Primary
Antigen: BCAR1 (breast cancer anti-estrogen resistance 1)
Clonality: monoclonal
Clone: EPR1860
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR