Anti-PCNA Rabbit Monoclonal Antibody [clone: EPR3821]
Lieferant: Abcam
ABCAAB92552-100EA 954 EUR
ABCAAB92552-100
Anti-PCNA Rabbit Monoclonal Antibody [clone: EPR3821]
Antikörper
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone: EPR3821
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen