Anti-GCG Rabbit monoclonal antibody unconjugated [clone: EP3070]

Lieferant: Abcam
ABCAAB92517-100EA 943 EUR
ABCAAB92517-100
Anti-GCG Rabbit monoclonal antibody unconjugated [clone: EP3070]
Antikörper
Type: Primary
Antigen: GCG (glucagon)
Clonality: monoclonal
Clone: EP3070
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR