Anti-P2RY6 Rabbit Monoclonal Antibody [clone: EPR3816]

Lieferant: Abcam
ABCAAB92504-100EA 806 EUR
ABCAAB92504-100
Anti-P2RY6 Rabbit Monoclonal Antibody [clone: EPR3816]
Antikörper
Type: Primary
Antigen: P2RY6 (pyrimidinergic receptor P2Y, G-protein coupled, 6)
Clonality: monoclonal
Clone: EPR3816
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR