Anti-CRTC1 Rabbit monoclonal antibody unconjugated [clone: EPR3382]

Lieferant: Abcam
ABCAAB92477-100EA 787 EUR
ABCAAB92477-100
Anti-CRTC1 Rabbit monoclonal antibody unconjugated [clone: EPR3382]
Antikörper
Type: Primary
Antigen: CRTC1 (CREB regulated transcription coactivator 1)
Clonality: monoclonal
Clone: EPR3382
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR