Anti-SLC22A2 Rabbit Monoclonal Antibody [clone: EPR542]

Lieferant: Abcam
ABCAAB92458-100EA 914 EUR
ABCAAB92458-100
Anti-SLC22A2 Rabbit Monoclonal Antibody [clone: EPR542]
Antikörper
Type: Primary
Antigen: SLC22A2 (solute carrier family 22 (organic cation transporter), member 2)
Clonality: monoclonal
Clone: EPR542
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR