Anti-Creatine kinase B type Rabbit monoclonal antibody unconjugated [clone: EPR3927]

Lieferant: Abcam
ABCAAB92452-100EA 930 EUR
ABCAAB92452-100
Anti-Creatine kinase B type Rabbit monoclonal antibody unconjugated [clone: EPR3927]
Antikörper
Type: Primary
Antigen: Creatine kinase B type
Clonality: monoclonal
Clone: EPR3927
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR