Anti-CEND1 Rabbit monoclonal antibody unconjugated [clone: EPR3738]

Lieferant: Abcam
ABCAAB92318-100EA 758 EUR
ABCAAB92318-100
Anti-CEND1 Rabbit monoclonal antibody unconjugated [clone: EPR3738]
Antikörper
Type: Primary
Antigen: CEND1 (cell cycle exit and neuronal differentiation 1)
Clonality: monoclonal
Clone: EPR3738
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR