Anti-SLC47A1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB92295-100EA 840 EUR
ABCAAB92295-100
Anti-SLC47A1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC47A1 (solute carrier family 47 (multidrug and toxin extrusion), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR