Anti-TSPO Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB92291-100EA 840 EUR
ABCAAB92291-100
Anti-TSPO Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TSPO (translocator protein (18kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR