Anti-Swine Influenza A Hemagglutinin Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB91641-100EA 837 EUR
ABCAAB91641-100
Anti-Swine Influenza A Hemagglutinin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Swine Influenza A Hemagglutinin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR