Anti-TNNT2 Rabbit Monoclonal Antibody [clone: EPR3695]

Lieferant: Abcam
ABCAAB91605-100EA 852 EUR
ABCAAB91605-100
Anti-TNNT2 Rabbit Monoclonal Antibody [clone: EPR3695]
Antikörper
Type: Primary
Antigen: TNNT2 (troponin T type 2 (cardiac))
Clonality: monoclonal
Clone: EPR3695
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR