Anti-CD98 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: FG 1/8]

Lieferant: Abcam
ABCAAB91297-50EA 834 EUR
ABCAAB91297-50
Anti-CD98 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: FG 1/8]
Antikörper
Type: Primary
Antigen: CD98
Clonality: monoclonal
Clone: FG 1/8
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR