Anti-NOS3 Mouse Monoclonal Antibody [clone: 6H2]

Lieferant: Abcam
ABCAAB91205-100EA 859 EUR
ABCAAB91205-100
Anti-NOS3 Mouse Monoclonal Antibody [clone: 6H2]
Antikörper
Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: monoclonal
Clone: 6H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR