Anti-Glycophorin A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI264]
Lieferant: Abcam
ABCAAB91163-50EA 423 EUR
ABCAAB91163-50
Anti-Glycophorin A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI264]
Antikörper
Type: Primary
Antigen: Glycophorin A
Clonality: monoclonal
Clone: HI264
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human