Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 47-3D6]

Lieferant: Abcam
ABCAAB91144-50EA 423 EUR
ABCAAB91144-50
Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 47-3D6]
Antikörper
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: monoclonal
Clone: 47-3D6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR