Anti-CSAD Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB91016-100EA 792 EUR
ABCAAB91016-100
Anti-CSAD Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CSAD (cysteine sulfinic acid decarboxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR