Anti-IL20RA Mouse Monoclonal Antibody [clone: MM0394-9F40]

Lieferant: Abcam
ABCAAB90935-50EA 853 EUR
ABCAAB90935-50
Anti-IL20RA Mouse Monoclonal Antibody [clone: MM0394-9F40]
Antikörper
Type: Primary
Antigen: IL20RA (interleukin 20 receptor, alpha)
Clonality: monoclonal
Clone: MM0394-9F40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR