Anti-Prosurfactant Protein C Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB90716-100EA 885 EUR
ABCAAB90716-100
Anti-Prosurfactant Protein C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Prosurfactant Protein C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR