Anti-MAML2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB90592-100EA 853 EUR
ABCAAB90592-100
Anti-MAML2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAML2 (mastermind-like 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR