Anti-HMOX2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB90515-50EA 878 EUR
ABCAAB90515-50
Anti-HMOX2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HMOX2 (heme oxygenase (decycling) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR