Anti-ACTN Mouse monoclonal antibody [clone: 7A4]

Lieferant: Abcam
ABCAAB90421-100EA 755 EUR
ABCAAB90421-100
Anti-ACTN Mouse monoclonal antibody [clone: 7A4]
Type: Primary
Antigen: ACTN (actinin, alpha)
Clonality: monoclonal
Clone: 7A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR